Assay Details
| Gene Symbol: LOC101106410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963455.1 | XP_027819256.1 | 1 - 2 | 45 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101106410| Gene Symbol: LOC101106410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963455.1 | XP_027819256.1 | 1 - 2 | 45 | 64 | |
Set Membership: |
|