Assay Details
| Gene Symbol: ZAR1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973019.1 | XP_027828820.1 | 3 - 4 | 1286 | 74 | |
| XM_027973021.1 | XP_027828822.1 | 2 - 3 | 1193 | 74 | ||
More Information
Set Membership: |
|

ZAR1L
Genomic Map