Assay Details
| Gene Symbol: CRLF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015104672.2 | XP_014960158.2 | 3 - 4 | 363 | 73 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

CRLF2| Gene Symbol: CRLF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015104672.2 | XP_014960158.2 | 3 - 4 | 363 | 73 | |