Assay Details
| Gene Symbol: LOC101109482 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027979877.1 | XP_027835678.1 | 14 - 15 | 2657 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101109482| Gene Symbol: LOC101109482 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027979877.1 | XP_027835678.1 | 14 - 15 | 2657 | 82 | |
Set Membership: |
|