Assay Details
| Gene Symbol: PROX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015097183.2 | XP_014952669.2 | 2 - 3 | 1400 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PROX2| Gene Symbol: PROX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015097183.2 | XP_014952669.2 | 2 - 3 | 1400 | 88 | |
Set Membership: |
|