Assay Details
| Gene Symbol: CEACAM20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977938.1 | XP_027833739.1 | 6 - 7 | 1588 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CEACAM20| Gene Symbol: CEACAM20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977938.1 | XP_027833739.1 | 6 - 7 | 1588 | 59 | |
Set Membership: |
|