Assay Details
| Gene Symbol: SPART | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012120.4 | XP_004012169.2 | 4 - 5 | 1299 | 137 | |
| XM_015098118.2 | XP_014953604.2 | 4 - 5 | 1284 | 137 | ||
| XM_027973610.1 | XP_027829411.1 | 4 - 5 | 1231 | 137 | ||
More Information
Set Membership: |
|

SPART
Genomic Map