Assay Details
| Gene Symbol: LOC101117690 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004014882.4 | XP_004014931.4 | 4 - 5 | 555 | 93 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101117690| Gene Symbol: LOC101117690 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004014882.4 | XP_004014931.4 | 4 - 5 | 555 | 93 | |