Assay Details
| Gene Symbol: PANX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004019501.4 | XP_004019550.1 | 2 - 3 | 410 | 62 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

PANX3| Gene Symbol: PANX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004019501.4 | XP_004019550.1 | 2 - 3 | 410 | 62 | |