Assay Details
| Gene Symbol: CITED1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004022185.3 | XP_004022234.2 | 2 - 3 | 488 | 85 | |
| XM_004022187.4 | XP_004022236.2 | 2 - 3 | 259 | 85 | ||
| XM_012106916.3 | XP_011962306.2 | 2 - 3 | 217 | 85 | ||
| XM_027963168.1 | XP_027818969.1 | 3 - 4 | 237 | 85 | ||
| XM_027963169.1 | XP_027818970.1 | 3 - 4 | 228 | 85 | ||
| XM_027963170.1 | XP_027818971.1 | 2 - 3 | 214 | 85 | ||
More Information
Set Membership: |
|

CITED1
Genomic Map