Assay Details
Gene Symbol: FBXL18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027961636.1 | XP_027817437.1 | 4 | 9665 | 103 | |
XM_027961637.1 | XP_027817438.1 | 5 | 5914 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
