Assay Details
| Gene Symbol: LOC101109964 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012177858.3 | XP_012033248.2 | 2 - 3 | 455 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101109964| Gene Symbol: LOC101109964 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012177858.3 | XP_012033248.2 | 2 - 3 | 455 | 75 | |
Set Membership: |
|