Assay Details
| Gene Symbol: LOC101111664 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015101736.2 | XP_014957222.2 | 3 - 4 | 418 | 71 | |
| XM_027956837.1 | XP_027812638.1 | 3 - 4 | 417 | 71 | ||
| XM_027956838.1 | XP_027812639.1 | 3 - 4 | 518 | 71 | ||
| XM_027956840.1 | XP_027812641.1 | 4 - 5 | 585 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015101736.2, XM_027956838.1, XM_027956840.1 transcripts.
Set Membership: |
|

LOC101111664
Genomic Map