Assay Details
| Gene Symbol: SCML4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011230.4 | XP_004011279.1 | 6 - 7 | 1242 | 66 | |
| XM_012182674.3 | XP_012038064.1 | 2 - 3 | 922 | 66 | ||
| XM_027972391.1 | XP_027828192.1 | 4 - 5 | 1196 | 66 | ||
More Information
Set Membership: |
|

SCML4
Genomic Map