Assay Details
| Gene Symbol: ZNF34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973389.1 | XP_027829190.1 | 3 - 4 | 653 | 59 | |
| XM_027973390.1 | XP_027829191.1 | 3 - 4 | 662 | 59 | ||
| XM_027973391.1 | XP_027829192.1 | 4 - 5 | 489 | 59 | ||
| XM_027973392.1 | XP_027829193.1 | 3 - 4 | 436 | 59 | ||
| XM_027973393.1 | XP_027829194.1 | 4 - 5 | 397 | 59 | ||
| XM_027973394.1 | XP_027829195.1 | 2 - 3 | 2071 | 59 | ||
| XM_027973395.1 | XP_027829196.1 | 3 - 4 | 344 | 59 | ||
| XM_027973396.1 | XP_027829197.1 | 3 - 4 | 354 | 59 | ||
| XM_027973397.1 | XP_027829198.1 | 3 - 4 | 436 | 59 | ||
| XM_027973398.1 | XP_027829199.1 | 2 - 3 | 2163 | 59 | ||
| XM_027973399.1 | XP_027829200.1 | 3 - 4 | 344 | 59 | ||
| XM_027973400.1 | XP_027829201.1 | 3 - 4 | 354 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027973397.1, XM_027973398.1, XM_027973399.1, XM_027973400.1 transcripts.
Set Membership: |
|

ZNF34
Genomic Map