Assay Details
| Gene Symbol: LOC101119572 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012469.4 | XP_004012518.1 | 1 - 2 | 167 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101119572| Gene Symbol: LOC101119572 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012469.4 | XP_004012518.1 | 1 - 2 | 167 | 108 | |
Set Membership: |
|