Assay Details
| Gene Symbol: TP53RK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004014619.4 | XP_004014668.1 | 1 - 2 | 1632 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TP53RK| Gene Symbol: TP53RK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004014619.4 | XP_004014668.1 | 1 - 2 | 1632 | 108 | |
Set Membership: |
|