Assay Details
| Gene Symbol: PAFAH1B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012095505.3 | XP_011950895.1 | 3 - 4 | 281 | 85 | |
| XM_027979229.1 | XP_027835030.1 | 3 - 4 | 302 | 85 | ||
More Information
Set Membership: |
|

PAFAH1B2
Genomic Map