Assay Details
| Gene Symbol: LOC101114909 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018502.3 | XP_004018551.1 | 4 - 5 | 667 | 101 | |
| XM_012100149.2 | XP_011955539.1 | 4 - 5 | 1782 | 101 | ||
| XM_012100151.2 | XP_011955541.1 | 5 - 6 | 1375 | 101 | ||
| XM_012100154.2 | XP_011955544.1 | 4 - 5 | 1286 | 101 | ||
More Information
Set Membership: |
|

LOC101114909
Genomic Map