Assay Details
| Gene Symbol: LOC101122140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015102191.2 | XP_014957677.1 | 2 - 3 | 175 | 78 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101122140| Gene Symbol: LOC101122140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015102191.2 | XP_014957677.1 | 2 - 3 | 175 | 78 | |