Assay Details
| Gene Symbol: LOC101115658 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958154.1 | XP_027813955.1 | 31 - 32 | 3253 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101115658| Gene Symbol: LOC101115658 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958154.1 | XP_027813955.1 | 31 - 32 | 3253 | 71 | |
Set Membership: |
|