Assay Details
| Gene Symbol: GARRE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027978203.1 | XP_027834004.1 | 12 - 13 | 3789 | 104 | |
| XM_027978204.1 | XP_027834005.1 | 11 - 12 | 3542 | 104 | ||
| XM_027978205.1 | XP_027834006.1 | 11 - 12 | 3996 | 104 | ||
| XM_027978206.1 | XP_027834007.1 | 11 - 12 | 3707 | 104 | ||
More Information
Set Membership: |
|

GARRE1
Genomic Map