Assay Details
| Gene Symbol: LOC101121817 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960978.1 | XP_027816779.1 | 2 - 3 | 322 | 71 | |
| XM_027960979.1 | XP_027816780.1 | 2 - 3 | 322 | 71 | ||
| XM_027960980.1 | XP_027816781.1 | 2 - 3 | 322 | 71 | ||
| XM_027960981.1 | XP_027816782.1 | 2 - 3 | 322 | 71 | ||
| Gene Symbol: LOC114110477 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960982.1 | XP_027816783.1 | 2 - 3 | 356 | 71 | |
More Information
Set Membership: |
|

LOC101121817
Genomic Map