Assay Details
| Gene Symbol: LOC101122294 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012187835.3 | XP_012043225.2 | 12 - 13 | 1371 | 91 | |
| XM_012187837.3 | XP_012043227.2 | 11 - 12 | 1323 | 91 | ||
| XM_012187844.3 | XP_012043234.2 | 10 - 11 | 1242 | 91 | ||
| XM_012187847.3 | XP_012043237.2 | 9 - 10 | 1194 | 91 | ||

LOC101122294
Genomic Map