Assay Details
| Gene Symbol: LOC101122294 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012187835.3 | XP_012043225.2 | 5 - 6 | 865 | 97 | |
| XM_012187836.3 | XP_012043226.2 | 5 - 6 | 865 | 97 | ||
| XM_012187837.3 | XP_012043227.2 | 5 - 6 | 865 | 97 | ||
| XM_012187838.3 | XP_012043228.2 | 5 - 6 | 865 | 97 | ||
| XM_012187839.3 | XP_012043229.2 | 5 - 6 | 865 | 97 | ||
| XM_012187840.3 | XP_012043230.2 | 5 - 6 | 865 | 97 | ||
| XM_012187841.3 | XP_012043231.2 | 5 - 6 | 865 | 97 | ||
| XM_012187843.3 | XP_012043233.2 | 5 - 6 | 865 | 97 | ||
| XM_012187844.3 | XP_012043234.2 | 5 - 6 | 865 | 97 | ||
| XM_012187847.3 | XP_012043237.2 | 5 - 6 | 865 | 97 | ||
| XM_012187848.3 | XP_012043238.2 | 5 - 6 | 865 | 97 | ||
| XM_012187849.3 | XP_012043239.2 | 5 - 6 | 865 | 97 | ||
| XM_012187850.3 | XP_012043240.2 | 5 - 6 | 865 | 97 | ||
| XM_027975989.1 | XP_027831790.1 | 5 - 6 | 865 | 97 | ||
| XM_027975990.1 | XP_027831791.1 | 5 - 6 | 865 | 97 | ||
| GenBank mRNA | KJ561887.1 | - | 5 - 6 | 749 | 97 | |

LOC101122294
Genomic Map