Assay Details
| Gene Symbol: LOC101119640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012180096.2 | XP_012035486.2 | 1 - 2 | 746 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101119640| Gene Symbol: LOC101119640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012180096.2 | XP_012035486.2 | 1 - 2 | 746 | 94 | |
Set Membership: |
|