Assay Details
| Gene Symbol: LOC101122884 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977463.1 | XP_027833264.1 | 1 - 2 | 105 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101122884| Gene Symbol: LOC101122884 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977463.1 | XP_027833264.1 | 1 - 2 | 105 | 89 | |
Set Membership: |
|