Assay Details
Gene Symbol: C1H1orf162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002319.4 | XP_004002368.1 | 2 - 3 | 485 | 82 | |
XM_027973054.1 | XP_027828855.1 | 2 - 3 | 485 | 82 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C1H1orf162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002319.4 | XP_004002368.1 | 2 - 3 | 485 | 82 | |
XM_027973054.1 | XP_027828855.1 | 2 - 3 | 485 | 82 |