Assay Details
| Gene Symbol: C1H1orf162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002319.4 | XP_004002368.1 | 4 - 5 | 641 | 93 | |
| XM_027973054.1 | XP_027828855.1 | 4 - 5 | 641 | 93 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1H1orf162| Gene Symbol: C1H1orf162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002319.4 | XP_004002368.1 | 4 - 5 | 641 | 93 | |
| XM_027973054.1 | XP_027828855.1 | 4 - 5 | 641 | 93 | ||