Assay Details
| Gene Symbol: C1H3orf38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002845.4 | XP_004002894.2 | 1 - 2 | 455 | 107 | |
| XM_012187993.3 | XP_012043383.2 | 1 - 2 | 455 | 107 | ||
More Information
Set Membership: |
|

C1H3orf38
Genomic Map