Assay Details
| Gene Symbol: NCOA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012105545.2 | XP_011960935.2 | 5 - 6 | 615 | 84 | |
| XM_012105546.3 | XP_011960936.2 | 5 - 6 | 572 | 84 | ||
| XM_012105548.2 | XP_011960938.2 | 5 - 6 | 615 | 84 | ||
More Information
Set Membership: |
|

NCOA4
Genomic Map