Assay Details
| Gene Symbol: BRD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027968226.1 | XP_027824027.1 | 1 - 2 | 1875 | 59 | |
| XM_027968227.1 | XP_027824028.1 | 2 - 3 | 1411 | 59 | ||
| XM_027968228.1 | XP_027824029.1 | 1 - 2 | 1875 | 59 | ||
| XM_027968229.1 | XP_027824030.1 | 1 - 2 | 1875 | 59 | ||
| XM_027968230.1 | XP_027824031.1 | 1 - 2 | 1886 | 59 | ||
| XM_027968231.1 | XP_027824032.1 | 1 - 2 | 1886 | 59 | ||
More Information
Set Membership: |
|

BRD1
Genomic Map