Assay Details
| Gene Symbol: PROZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973815.1 | XP_027829616.1 | 4 - 5 | 576 | 63 | |
| XM_027973816.1 | XP_027829617.1 | 5 - 6 | 677 | 63 | ||
More Information
Set Membership: |
|

PROZ
Genomic Map