Assay Details
| Gene Symbol: FAM20C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961840.1 | XP_027817641.1 | 6 - 7 | 1415 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM20C| Gene Symbol: FAM20C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961840.1 | XP_027817641.1 | 6 - 7 | 1415 | 61 | |
Set Membership: |
|