Assay Details
| Gene Symbol: TRMU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027968216.1 | XP_027824017.1 | 2 - 3 | 311 | 101 | |
| XM_027968217.1 | XP_027824018.1 | 2 - 3 | 346 | 101 | ||
| XM_027968218.1 | XP_027824019.1 | 2 - 3 | 302 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027968218.1 transcripts.
Set Membership: |
|

TRMU
Genomic Map