Assay Details
| Gene Symbol: LOC105611052 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975421.1 | XP_027831222.1 | 7 - 8 | 619 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105611052| Gene Symbol: LOC105611052 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975421.1 | XP_027831222.1 | 7 - 8 | 619 | 77 | |
Set Membership: |
|