Assay Details
| Gene Symbol: LOC114112279 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963771.1 | XP_027819572.1 | 3 - 4 | 422 | 110 | |
| Gene Symbol: OBSCN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969216.1 | XP_027825017.1 | 33 - 34 | 8099 | 110 | |
More Information
Set Membership: |
|

LOC114112279
Genomic Map