Assay Details
| Gene Symbol: LOC114112279 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963771.1 | XP_027819572.1 | 6 - 7 | 1230 | 104 | |
| Gene Symbol: OBSCN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969216.1 | XP_027825017.1 | 36 - 37 | 8907 | 104 | |
More Information
Set Membership: |
|

LOC114112279
Genomic Map