Assay Details
| Gene Symbol: CEBPZOS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027966925.1 | XP_027822726.1 | 2 - 3 | 138 | 71 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

CEBPZOS| Gene Symbol: CEBPZOS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027966925.1 | XP_027822726.1 | 2 - 3 | 138 | 71 | |