Assay Details
| Gene Symbol: CEBPZOS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027966926.1 | XP_027822727.1 | 3 - 4 | 245 | 100 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

CEBPZOS| Gene Symbol: CEBPZOS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027966926.1 | XP_027822727.1 | 3 - 4 | 245 | 100 | |