Assay Details
| Gene Symbol: DNASE1L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027962029.1 | XP_027817830.1 | 2 - 3 | 1298 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DNASE1L2| Gene Symbol: DNASE1L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027962029.1 | XP_027817830.1 | 2 - 3 | 1298 | 66 | |
Set Membership: |
|