Assay Details
| Gene Symbol: RBM23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010346.4 | XP_004010395.1 | 10 - 11 | 1186 | 59 | |
| XM_012180994.3 | XP_012036384.1 | 9 - 10 | 1620 | 59 | ||
| XM_012180997.3 | XP_012036387.1 | 12 - 13 | 1270 | 59 | ||
| XM_012180998.3 | XP_012036388.1 | 13 - 14 | 1292 | 59 | ||
| XM_012181001.3 | XP_012036391.1 | 11 - 12 | 1217 | 59 | ||
| XM_012181002.3 | XP_012036392.1 | 12 - 13 | 1239 | 59 | ||
| XM_015096885.2 | XP_014952371.1 | 12 - 13 | 2411 | 59 | ||
| XM_027971645.1 | XP_027827446.1 | 12 - 13 | 3419 | 59 | ||
| XM_027971646.1 | XP_027827447.1 | 11 - 12 | 2358 | 59 | ||
More Information
Set Membership: |
|

RBM23
Genomic Map