Assay Details
| Gene Symbol: MRM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004013238.4 | XP_004013287.2 | 3 - 4 | 750 | 113 | |
| XM_027975164.1 | XP_027830965.1 | 2 - 3 | 254 | 113 | ||
More Information
Set Membership: |
|

MRM3
Genomic Map