Assay Details
| Gene Symbol: ADSS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027957466.1 | XP_027813267.1 | 9 - 10 | 1012 | 88 | |
| XM_027957467.1 | XP_027813268.1 | 9 - 10 | 1038 | 88 | ||
More Information
Set Membership: |
|

ADSS1
Genomic Map