Assay Details
| Gene Symbol: MFGE8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027957063.1 | XP_027812864.1 | 7 - 8 | 1097 | 62 | |
| GenBank mRNA | JN016710.1 | - | 6 - 7 | 912 | 62 | |
More Information
Set Membership: |
|

MFGE8
Genomic Map