Assay Details
| Gene Symbol: HIVEP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972502.1 | XP_027828303.1 | 4 - 5 | 5795 | 83 | |
| XM_027972503.1 | XP_027828304.1 | 4 - 5 | 5795 | 83 | ||
More Information
Set Membership: |
|

HIVEP2
Genomic Map