Assay Details
| Gene Symbol: LOC554335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974354.1 | XP_027830155.1 | 14 - 15 | 2226 | 100 | |
| XM_027974355.1 | XP_027830156.1 | 8 - 9 | 1192 | 100 | ||
| XM_027974356.1 | XP_027830157.1 | 3 - 4 | 836 | 100 | ||
| GenBank mRNA | AJ920032.1 | - | 3 - 4 | 468 | 100 | |
More Information
Set Membership: |
|

LOC554335
Genomic Map