Assay Details
| Gene Symbol: LOC114117808 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977572.1 | XP_027833373.1 | 2 - 3 | 430 | 79 | |
| XM_027977575.1 | XP_027833376.1 | 1 - 2 | 337 | 79 | ||
| XM_027977579.1 | XP_027833380.1 | 2 - 3 | 430 | 79 | ||
| Gene Symbol: UAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977559.1 | XP_027833360.1 | 2 - 3 | 480 | 79 | |
| XM_027977565.1 | XP_027833366.1 | 3 - 4 | 685 | 79 | ||
| XM_027977570.1 | XP_027833371.1 | 2 - 3 | 480 | 79 | ||

LOC114117808
Genomic Map