Assay Details
| Gene Symbol: AK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004005007.3 | XP_004005056.2 | 2 - 3 | 290 | 61 | |
| XM_012148993.2 | XP_012004383.2 | 2 - 3 | 307 | 61 | ||
| XM_012149013.2 | XP_012004403.1 | 2 - 3 | 355 | 61 | ||
| XM_027965328.1 | XP_027821129.1 | 2 - 3 | 406 | 61 | ||
| XM_027965329.1 | XP_027821130.1 | 2 - 3 | 294 | 61 | ||
More Information
Set Membership: |
|

AK2
Genomic Map