Assay Details
| Gene Symbol: LOC101108212 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027967845.1 | XP_027823646.1 | 24 - 25 | 3406 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101108212| Gene Symbol: LOC101108212 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027967845.1 | XP_027823646.1 | 24 - 25 | 3406 | 57 | |
Set Membership: |
|